Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AlphaTec® Neoprene Isolator Gloves, 55-308 Series, Ansell

Supplier: Ansell
AlphaTec®
55308100
76413-640CS 4618.96 USD InStock
76413-640
AlphaTec® Neoprene Isolator Gloves, 55-308 Series, Ansell
Gloves
Non-clean/non sterile neoprene RABS/Isolator gloves for use in 305 mm (12") ports.

  • Effective protection for heavy duty chemical environments
  • Port size: 305 mm (12")
  • Exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons
  • Heavyweight thickness (30 mil/0.76 mm) increases durability, augmenting chemical and physical resistance
  • Unlined for effective cleaning

AlphaTec® Neoprene RABS and Isolator gloves deliver exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons. They offer strong physical protection against abrasions and good protection against cuts and punctures.

Certifications: CE 0493, EN 374:2003, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN 388:2016, EN 420:2003 + A1:2009, Category III, UKCA 0321.

Packaging: 1 pair per black sealed plastic bag; 10 pairs per shipper carton.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty