Would you like to log in again?
RCT 5 DIGITAL MAGNETIC STIRRER 850 W
Supplier: IKA WORKS
RCT 5 digital, magnetic stirrer, with 850 W offers significantly more power making it an ideal device for demanding stirring tasks up to 20 l, Scratch-resistant ceramic coating, rectangular set-up plate with a white, safety circuit can be adjusted up to 370 C, less scratch-prone
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RCT 5 DIGITAL MAGNETIC STIRRER 850 W 76414-088 0020002705 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty