Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IKAMAG® RET Basic Safety Control, IKA

Supplier: IKA
IKAMAG®
0003622001
76414-090EA 1814.7 USD InStock
76414-090
IKAMAG® RET Basic Safety Control, IKA
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrer with extremely fast heating times and integrated temperature control, suitable for unsupervised operation.

  • Exact temperature and speed setting via digital display, even when switched off
  • Safety temperature limit that has been set is displayed digitally
  • Increased safety due to improved heat control technology, hot warning light helps to prevent burns
  • Adjustable safety circuit of heating plate temperature (50 to 380 °C)
  • Can be used at 5 to 40 °C (80% relative humidity)
  • LED speed display

Units have bushing according to DIN 12878 for connecting a contact thermometer, enabling precise temperature control. Features include high magnetic adhesion, electronic speed control and digital error code display.

Maximum stirring bar length of 80 mm, Heating rate of 7 K/min.

Certifications: IP Protection Class according to DIN EN 60529: IP 42

Delivery information: Supplied with PT1000 temperature sensor and silicone protective cover, accessories must be ordered separately.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty