Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lab disc White Magnetic Stirrers, IKA Works
Supplier: IKA
0003907500
76414-092EA 493.35 USD InStock
76414-092
Lab disc White Magnetic Stirrers, IKA Works
Stirrers Magnetic Stirrers
Ultra-flat magnetic stirrers with magnetic coil technology. Top plate and casing are made of chemically resistant materials. Wear-free drive with no moving parts.
  • Reverses direction of rotation every 30 seconds to ensure optimum mixing (selectable)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
Stepless speed adjustment.
Certifications: IP protection class according to DIN EN 60529: IP 65
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty