Would you like to log in again?
MIDI MR 1 DIGITAL MAGNETIC STIRRER
Supplier: IKA WORKS
Midi MR 1 digital, Powerful magnetic stirrer without heating function, Stirring quantity up to 50 liters (H2O), Flat, sturdy stainless steel casing, Non-locking motor, Infinitely variable speed, Digital LED speed display, Timer (0 - 56 min) or continuous operation, Integrated USB, RS 232 interface
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MIDI MR 1 DIGITAL MAGNETIC STIRRER 76414-094 0025002970 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty