Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Digital Mechanical Overhead Stirrer, RW 20, IKA® Mixing

Supplier: IKA
0003593001
76414-100EA 1719.7 USD InStock
76414-100
Digital Mechanical Overhead Stirrer, RW 20, IKA® Mixing
Stirrers Overhead Stirrers
RW 20 mechanical overhead stirrer is robust, slimline, ergonomic design with digital display.

  • With constant power-drive; stepless speed control
  • Two speed ranges for universal use from 60 to 2000 rpm
  • Push through agitator shafts (only when stationary)

Technical improvements on the trusted RW 20 series designs.

Output shaft: 26 W

Certifications: IP protection class according to DIN EN 60529: IP 20

Ordering information: A wide range of accessories are available to order separately.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty