Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Reciprocating Shakers, HS 501 digital

Supplier: IKA
0025002983
76414-118EA 5810.29 USD InStock
76414-118
Reciprocating Shakers, HS 501 digital
Shakers and Mixers Reciprocating Shakers
Low profile laboratory shaker with digital display and a large platform.

  • Infinitely variable speed control
  • Continuous operation or timer operation from 0 to 56 min
  • Electronic speed control
  • Can be used at 5 to 50 °C (80% relative humidity), in an incubator or cold room

Continuous operation is guaranteed even under extreme loads. Suitable for vessels which need to be laid flat, for example separating funnels. A variety of different vessels can be accommodated by choosing from the range of optional holding attachments.

Certifications: IP protection class according to DIN EN 60529: IP 21

Accessories information: Shaker HS 501 digital accommodates a maximum of  50×25, 48×50, 25×100, 16×250 or 12×500 ml flasks.

Ordering information: Supplied without attachments, required accessories must be ordered separately.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty