Would you like to log in again?
RC 5 BASIC TEMPERATURE CTRL 1400W -30C
Supplier: IKA WORKS
RC 5 basic, Temperature Control, Powerful recirculating chiller, designed for external cooling applications up to -30 deg C, Speed-controlled refrigeration system with 1400 W cooling capacity (20C), Natural thermofluid R290, extended operating temperature range until 80C, Digital level indicator
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RC 5 BASIC TEMPERATURE CTRL 1400W -30C 76414-128 0020008503 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty