Would you like to log in again?
FILTER CARTRIDGE XL SERIES SI 5UM 30IN
Supplier: 3M OCCUPATIONAL HEALTH
Betafine* XL Filter Cartridge Series, absolute-rated, made of a 100% polypropylene pleated filter featuring Advanced Pleat Technology (APT), 226 O-ring & Spear, No Reinforcing Ring, Silicone Gasket/O-ring Material, Micron Rating: 5um, reduce quality checks, product rejects, Overall Length: 30 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER CARTRIDGE XL SERIES SI 5UM 30IN 76414-142 7100006948 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty