Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Emricasan ≥98% (by HPLC)

Warning
Synonyms: (S)-3-((S)-2-(2-((2-(tert-Butyl)phenyl)amino)-2-oxoacetamido)propanamido)-4-oxo-5-(2,3,5,6-tetrafluorophenoxy)pentanoic acid

AB287102-5MG AB287102-25MG
76414-578EA 194.96 USD InStock
76414-578 76414-576
Emricasan ≥98% (by HPLC)
Emricasan
Emricasan (PF-03491390) is an irreversible pan-caspase inhibitor. In a mouse model of α-Fas-induced acute liver injury, the orally-administered caspase inhibitor Emricasan was retained in the liver for prolonged periods with a low systemic exposure.

  • A caspase inhibitor


Formula: C₂₆H₂₇F₄N₃O₇
MW: 569.50 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11840420
CAS Number: 254750-02-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty