Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Azure Aqua Quad Mini-Cell
Supplier: Azure Biosystems
AC4200
76414-790EA 1092.79 USD InStock
76414-790
Azure Aqua Quad Mini-Cell
Electrophoresis Reagents Western Blotting Reagents
Compatible with mini gels cassette dimensions 10×8 cm.
  • Comes with 1 mm gel plate set
  • For use with polyacrylamide gels
  • Comprised of an upper and a lower buffer chamber
W×D×H: 14×10×15 cm
Ordering information: It includes 4 gel casting stands and casting frames, 1 buffer dam, 5 spacer plates (1mm), 10 short glass plates, 5 15-well combs, 5 10-well combs, 5 sealing gaskets, 5 gel shovels, Tank and upper lid with power cables, primary and secondary electrode cores.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty