Would you like to log in again?
CF® Dye BCN, Biotium
Supplier: Biotium
96026 92113 92114 92076 96070 96059 92075 92079 92077 96027 92078 96058
10001-656EA 419.72 USD InStock
10001-656 10001-658 76404-070 10066-550 10066-552 76221-524 76414-982 10066-554 76221-526 76414-980 10066-546 10066-548
CF® Dye BCN, Biotium
Protein Linking and Labeling Systems
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labeling reactions are also available.
  • Fluorescent labeling of azide groups on target molecules via copper-free bioorthonal chemistry
  • Alternative to fluorescently labeled DIBO and DBCO
  • Suitable for surface or intracellular labeling of live cells or cell extracts
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
  • Available in several CF® Dye options including membrane-permeant forms for intracellular labeling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labeled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labeling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labeling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty