Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CompactDry™ E. coli (EC), Hardy Diagnostics
Supplier: Hardy Diagnostics
CompactDry™
54082 54082
76414-990EA 235.48 USD InStock
76414-990 CA10145-954
CompactDry™ E. coli (EC), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ EC is a ready-to-use test method recommended for the isolation and enumeration of coliforms and Escherichia coli in raw materials, finished products, or on environmental surfaces pertaining to food and related industries.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
CompactDry™ EC contains two kinds of chromogenic enzyme substrates: Magenta-Gal and X-Gluc. E. coli forms blue colonies, coliforms turn pink-purple. The total coliform group count is the sum of both the pink-purple and blue colonies.
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty