Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

WypAll® Reusable Foodservice Cloths, Extended Use, Kimberly-Clark Professional

Supplier: Kimberly-Clark
WYPALL*
51639 51636
76415-176CS 57.05 USD InStock
76415-176 76415-174
WypAll® Reusable Foodservice Cloths, Extended Use, Kimberly-Clark Professional
Wipes Reusable Cleaning Cloths Cloths for General Applications
WypAll® extended use foodservice cloths are a hygienic alternative to either laundered towels or cloth rags for foodservice prep.

  • Durable, absorbent cloths stay strong for cleaning and sanitizing
  • Cleans residue that rags leave behind
  • Absorbs 25% faster than competitive disposable wipers
  • 200 quarter-folded sheets per box
  • Each foodservice cloth measures 12.5×23.5" (unfolded)

These cloths made with high-tech HydroKnit fast-absorbing material and suitable for all common cleaning and sanitizing tasks in foodservice industries.

WypAll® extended use foodservice cloths are strong enough for extended use, and can be machine washed, rinsed, and reused. Plus, they come conveniently and compactly packaged in a quarter-fold format (an alternative to a box of rags) and are so durable that they can often be used more than once. After using them a few times, they’re disposable.

Certifications: NSF and FSC certified.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty