Would you like to log in again?
GLOVES NEOPRENE 11.6IN CREAM SZ8.5 CS200
Supplier: Ansell Healthcare
TouchNTuff* Gloves, Clean Disposable, Material: Neoprene (Polychloroprene), Compatible with Class 100 (ISO 5) Environments, Thin design for superior tactile sensitivity, Smooth Surface, Prevents Type I and Type IV allergies, Color: Creamy, Palm Thickness: 5.90-5.91 mil, Length: 11.6in, Size: 8.5
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NEOPRENE 11.6IN CREAM SZ8.5 CS200 76415-412 73300085 Case of 200 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty