Would you like to log in again?
SAMPLE HOLDER F/FILM AQUAMATE 7100/8100
Supplier: Thermo Scientific
Sample holder, assembled in a removable sample compartment liner, for film/filter for Aquamate 7100/8100 Spectrophotometer, Holds Filters/lenses up to 50mm long, 80mm tall and 6mm thick, Includes: 1 Film / Filter Holder
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPLE HOLDER F/FILM AQUAMATE 7100/8100 76415-504 AQX1FLTRHDR Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty