Would you like to log in again?
CELL HOLDER LONG PATH RECTANGULAR F/7100
Supplier: Thermo Scientific
Long path rectangular cell holder assembled in a removable compartment liner, for AquaMate 7100 & 8100 Spectrophotometers, Accommodates 20-100 mm path length
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CELL HOLDER LONG PATH RECTANGULAR F/7100 76415-510 AQX1LWLVH Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty