Would you like to log in again?
Advanced Vortex Mixers, ZX4, VELP Scientifica
Supplier: VELP Scientifica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SA202A0280 F202A0280
76416-206EA 417.61 USD InStock
76416-206 76416-292
Advanced Vortex Mixers, ZX4, VELP Scientifica
Shakers and Mixers Vortex Mixers
Vortex mixer with adjustable stirring speed, with continuous and IR sensor operating mode.
  • Variable speed operation (electronically controlled)
  • Large choice of accessories to suit tubes, plates and flasks
  • Universal voltage/frequency model
  • Accessories fit by simple pressure
  • Four large, anti-slip rubber feet for staying in required position
Optimum resistance to chemicals from technopolymer casing, excellent stability with zinc alloy base.
The infrared sensor technology ensures a drastic reduction in repetitive strain.
Certifications: IP Protection Class according to CEI EN 60259: IP 42 protection against particles and liquids.
Delivery information: Three year warranty.
Supplied with small rubber supporting plate, customizable soft foam top and stand are available as options.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty