Would you like to log in again?
MULTISTIRRER Multiple Positions Magnetic Stirrers, Velp Scientific
Supplier: VELP Scientifica
F203A0177 F203A0178
76416-212EA 1948.09 USD InStock
76416-212 76416-210
MULTISTIRRER Multiple Positions Magnetic Stirrers, Velp Scientific
Stirrers Magnetic Stirrers, Multi-position
Multi-position magnetic stirrer with 6 or 15 positions, that remains cold even after several days of continuous use.
  • Control panel is shielded from liquid spills
  • Epoxy-coated tough steel casing
  • Stainless steal cover
  • Ergonomic - very low height
  • Speed regulation from 80 to 1500 rpm
  • Universal mains voltage/frequency range
  • Light weight, easy to move
Remains cold, even after days of continuous use. Pale surface for visualization of particles and color changes. Soft start but rapidly reaching set speed, excellent speed control (electronic), even at low speeds. Can thermostat samples using a recirculating water bath. Ideal for microtitration, microbiology and biochemistry.
Stirring system has high-power driving magnet type ''PCM'' operated by a mono-phase motor for continuous operation. Distance between stirring position centers for MULTISTIRRER 6 and MULTISTIRRER 15 is 100 and 74 mm.
Delivery information: Three year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty