Would you like to log in again?
AMI Illuminated Magnetic Stirrers, Velp Scientific
Supplier: VELP Scientifica
F204A0168
76416-232EA 2124.16 USD InStock
76416-232
AMI Illuminated Magnetic Stirrers, Velp Scientific
Stirrers Magnetic Stirrers, Multi-position
4-position illuminated magnetic stirrer, particularly useful for titrations.
  • Control panel is shielded from liquid spills
  • Epoxy-coated tough steel casing
  • Soft start but rapidly reaching set speed
  • Excellent speed control (electronic), even at low speed
  • Possibility to adjust the illumination brightness
  • Ergonomic - very low height
  • Universal mains voltage range, light weight
Illuminated surface for visualization of particles and color changes. Ideal for microtitration, in particular for subtle color changes. The four-position model provides independent speed regulation. High-power driving magnet type 'PCM' operated by a mono-phase motor for continuous operation stirring system.
Distance between stirring position centers of AMI 4 is 150 mm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty