Would you like to log in again?
Human GAPDH Inhibitor Screening Kit (Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB283369-100TEST
76416-480EA 1125.02 USD InStock
76416-480
Human GAPDH Inhibitor Screening Kit (Colorimetric), BioVision
Assays Enzyme Assays
To screen potential inhibitors of Human GAPDH
  • Simple
  • Sensitive
  • Quick and easy method for screening potential inhibitors of GAPDH
Glucose 3-Phosphate Dehydrogenase (GAPDH; EC 1.2.1.12) is a cytosolic enzyme, which catalyzes the conversion of Glyceraldehyde 3-Phosphate to 1,3-Bisphosphoglycerate (BPG) with the reduction of NAD to NADH in the glycolytic pathway. GAPDH is a multifunctional protein involved in many intracellular processes such as apoptosis, membrane trafficking, ion metabolism, nuclear translocation etc. Although GAPDH is considered as housekeeping gene, increased GAPDH activity has been detected in various human cancers. Additionally, knockdown of GAPDH has been shown to decrease cell proliferation, cell migration and metastasis in in vitro experiments. Therefore, targeting this key enzyme is essential for developing novel therapeutics for treating cancer. In our GAPDH Inhibitor Screening Kit, Glyceraldehyde 3-Phosphate is oxidized by GAPDH to generate 1,3-Bisphosphoglycerate and NADH, which reduces the probe thereby generating a strong absorbance at 450 nm. In the presence of GAPDH inhibitors, the reactions are impeded, thus decreasing the rate and/or extent of generation of GAPDH-dependent absorbance at OD 450 nm. This kit provides a sensitive, quick, and easy method for screening potential inhibitors of GAPDH. GAPDH Inhibitor Control is included in the kit to compare the efficacy of test inhibitors. The assay is high-throughput adaptable and can be performed in less than 30 min.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty