Would you like to log in again?
Social Distance Quik Sign Fold-Ups®; Safety First, Accuform®
Supplier: Accuform
PFK900
76417-056EA 227.63 USD InStock
76417-056
Social Distance Quik Sign Fold-Ups®; Safety First, Accuform®
Signs
Floor signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus. 
  • Molded polyethylene plastic stand is 30½H×26"W with feet 22" apart
  • Includes one 18H×24"W corrugated plastic printed sign that fits into slot
  • Handle holes for easy portability
Stand closes to 2" thick with molded-in lug points for nesting into each other when stacked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty