Would you like to log in again?
FACE MASK WASH CLOTH 100% COTTON CS600
Supplier: Avantor
Face Mask^ Face Mask, Machine wash, up to 15 times, 100% Cotton with antimicrobial finish, Covers nose, mouth and chin, Soft ear loops for a comfortable fit, Masks are designed to meet CDC recommendations, Double Layer Cover For Dust Particle & Droplet Protection
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE MASK WASH CLOTH 100% COTTON CS600 76417-114   Case of 600 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty