Would you like to log in again?
ReFIBE Wipes, Contec®
Supplier: Contec
RFHS-1212IR RFHS-99 RFHS-99 RFHS-1212 RFHS-1212 RFHS-99IR
76468-272CS 827.47 USD InStock
76468-272 76417-116 CA76417-116 76417-118 CA76417-118 76468-274
ReFIBE Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ReFIBE wipes are made from 100% recycled plastic bottles, making them a sustainable single-use option for critical manufacturers in the life sciences, pharmaceutical, medical device, and semi-conductor industries. The revolutionary recycling process converts post-consumer plastic to 100% continuous filament polyester, the same substrate used in many of standard polyester wipes and mops.
  • Made from recycled plastic bottles, diverting waste from landfills
  • 100% synthetic polyester construction is low in extractables and compatible with a wide variety of solvents
  • Low in particles and fibers, lowering the risk of contamination during use
  • Laser cut edge to be suitable for use in ISO 3 and higher environments
As synthetic polyester, they are exceptionally clean and are compatible with a wide range of cleanroom solutions and disinfectants. Low in releasable particles and fibers, they can be used in place of traditional polyester to incorporate sustainability without compromising quality or performance. Available in 9×9" and 12×12" sizes.
Sterile ReFIBE wipes have been validated sterile to an SAL of 10⁻⁶.
How wipes are made from recycled materials:
Post consumer bottles are collected
Bottles are chopped into flake and cleaned
Bottle flake is melted, filtered and formed into chip
Chip is melted and made into yarn
Yarn is knitted into ReFIBE wipes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty