Would you like to log in again?
Stainless Steel Multi-Bucket System, Contec®
Supplier: Contec
2707 2995 2617
76417-128EA 1209.71 USD InStock
76417-128 33503-124 76417-130
Stainless Steel Multi-Bucket System, Contec®
Mops
Stainless steel cart with two buckets and downpress wringer.
  • Includes two 27 L (7 gal.) buckets and one cart with four non-marking, autoclavable casters
  • Cart dimensions: 66×61×92 cm (26L×24W×36" H), with handle and casters
  • Buckets, wringer, and cart are manufactured from 304 electropolished stainless steel and are fully autoclavable
  • Buckets are deep drawn and seamless, reducing the risk of microbial contamination
  • A lip around the top edge of the buckets allows for easy lifting
The Stainless steel multi-bucket system offers maximum flexibility and can be used with two or three 27 L (7 gal.), depending upon in-house protocols. An extendable frame pulls out of the front of the cart to accommodate an additional bucket. Lightweight and easy to maneuver, the cart is fully autoclavable and features non-marring wheels. The stainless steel downpress wringer is compatible with all sizes of VertiKlean Mops and Edgeless String Mops.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty