Would you like to log in again?
VWR® Screw Cap Microtubes
Supplier: VWR
VWR®
2230-00-200AV 2220-00-200AV 2120-00-200AV 2241-S0-581AV 2320-00-200AV 2240-00-200AV 2141-S0-581AV 2341-S0-581AV 2121-S0-581AV 2530U00-200AV 2310-00-200AV 2640U00-200AV 2231-S0-581AV 2331-S0-581AV 2630U00-200AV 2340-00-200AV 2330-00-200AV 2140-00-200AV 2130-00-200AV 2210-00-200AV
76417-250CS 577.21 USD InStock
76417-250 76417-252 76417-240 76417-232 76417-210 76417-254 76417-242 76417-212 76417-256 76417-234 76417-244 76417-214 76417-258 76417-236 76417-246 76417-238 76417-204 76417-248 76417-206 76417-208
VWR® Screw Cap Microtubes
Tubes Microtubes
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
  • All tubes are manufactured from ultra-pure PP
  • High-clarity tubes provide excellent visibility to the sample
  • O-ringed flat caps feature a ³/₄ turn to open or close
  • Wide temperature range to meet a wide variety of applications: −196 to +121 °C, but not suitable for immersion in liquid nitrogen
  • Sterile assemblies or handy, non sterile combo packs are available
Screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty