Would you like to log in again?
Guardian 5000 Hotplates, OHAUS
Supplier: Ohaus
30500533 30500530
76417-278EA 359.62 USD InStock
76417-278 76417-280
Guardian 5000 Hotplates, OHAUS
Heaters Hot Plates
Reliable hotplates with exceptional safety features for everyday use. Guardian 5000 hotplates are designed for safety and simplicity in all research, academic and industrial applications.
  • SafetyHeat™ system protects the laboratory
  • Easy to read LCD display
  • Designed for durability and reliability with SmartHousing™
The design ensures temperature control of the top plate or your sample when using the optional temperature probe. Heating models include a Single Point Calibration feature for multiple setpoints.
The early detection system uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
The bright LCD display includes large temperature settings, intuitive icons and a prominent hot top indicator which are all designed to quickly and easily monitor performance from across the laboratory.
The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internal components and the angled control panel.
Corresponding magnetic hotplate stirrers (heating and stirring 76417-274 and 76417-286) or magnetic stirrers (stirring only 76417-282) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: Product Safety: TÜV SÜD certified to CAN/CSA C22.2 61010-1; CAN/CSA C22.2 61010- 2-010; CAN/CSA C22.2 61010-2-051; UL 61010-1; UL 61010-2-010; UL 61010-2-051
Electromagnetic Compatibility: Canada ICES-003 Class A; FCC Part 15 Class A
Compliance Mark: TÜV SÜD
Accessories information: Built-in support rod holder.
Ordering information: Two year manufacturer's warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty