Would you like to log in again?
Guardian 7000 Hotplate Stirrers, OHAUS
Supplier: Ohaus
30500610 30500600 30500620
76417-294EA 1021.76 USD InStock
76417-294 76417-296 76417-298
Guardian 7000 Hotplate Stirrers, OHAUS
Stirrers Magnetic Hotplate Stirrers
Durable hotplate stirrers with superior safety features and intelligent performance. These Guardian 7000 hotplate stirrers are designed with outstanding safety features and superior heating and mixing performance.
  • SmartPresence™ and SmartLink™ Technologies ensure supervised operation
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Customized performance for precise applications
SmartPresence™ and SmartLink™ technologies help protect the laboratory, they automatically shut off the heater if no one is detected after a user defined 'time out' period has passed. The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internal components and the angled glass control panel. The bright LCD display includes intuitive icons, temperature, speed and time settings which are all designed to monitor performance from across the laboratory.
An early detection system uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
SmartHeat™ allows you to set the maximum temperature of the unit and SmartRate™ enables fast or slow temperature and speed ramp rates. The programming feature allows storage of five multi-step programs.
Certifications: Product Safety: TÜV SÜD certified to CAN/CSA C22.2 61010-1; CAN/CSA C22.2 61010- 2-010; CAN/CSA C22.2 61010-2-051; UL 61010-1; UL 61010-2-010; UL 61010-2-051
Electromagnetic Compatibility: Canada ICES-003 Class A; FCC Part 15 Class A
Compliance Mark: TÜV SÜD
Accessories information: Built-in support rod holder, PTFE stir-bar.
Ordering information: Two year manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty