Would you like to log in again?
WEIGHT M1 STANDARD W/PLASTIC CASE 2KG
Supplier: Mettler Toledo
Weight^ Standard Weight, OIML Class: M1 E, without Calibration Certificate, with an adjusting cavity and a plastic box, Material: 304 Stainless steel, Density p: 7950 (+/- 140) kg/m3, Susceptibility X: < 0.8, for regular routine testing without the need for traceability, Nominal Value: 2kg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WEIGHT M1 STANDARD W/PLASTIC CASE 2KG 76417-480 30402665 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty