Would you like to log in again?
WEIGHT SET M1 W/PLASTIC CASE 1MG-2KG
Supplier: Mettler Toledo
Weight set^ Weight set, Standard, OIML Class: M1 E, in a plastic box including accessories for handling and cleaning, without Calibration Certificate, Density: 7950 (+/- 140) kg/m3, Material: 304 Stainless steel, for regular testing without the need for traceability, Content: 1mg-2kg(27 pieces)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WEIGHT SET M1 W/PLASTIC CASE 1MG-2KG 76417-502 30402682 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty