Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Vortex Mixers, Thermo Scientific

Supplier: Thermo Scientific
88882009
76417-524EA 532.48 USD InStock
76417-524
Digital Vortex Mixers, Thermo Scientific
Shakers and Mixers Vortex Mixers
Digital vortex mixer designed for accurate speed control when continuously mixing or pulsing samples.

  • Digital LED display of speed and time
  • Continuous and touch modes
  • Speed range: 200 to 3000 rpm
  • Precise speed control: ±5% rpm
  • Time set: up to 99 minutes

Two timer modes with Down-counting for repeat tests and continuous timer, time set as 0.

Quick set up to maximum rpm:
3 seconds 1500 rpm (Touch)
6 seconds 3000 rpm (Continuous)

Certifications: cCSAus, CE, RoHS.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty