Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Rockers, 230 V

Supplier: Thermo Scientific
88882002
76417-530EA 1344.45 USD InStock
76417-530
Digital Rockers, 230 V
Shakers and Mixers Rotating/Rocking Shakers
Digital Rocker is designed for ELISA, protein synthesis, gel staining and de-staining, antibody staining, and other time-sensitive applications.

  • Digital display of speed and time
  • Platform style: Tray
  • Capacity: 6×96-well plates
  • Timer range: Continuous or 1 s to 9999 minutes

Digital display with big platform, accurate speed control and quick angle adjustment for easy sample agitation. Optional non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags.

Certifications: cCSAus, CE, RoHS.

Ordering information: Two year warranty.

Delivery information: Includes dimpled rubber mat tray, rubber strip, user manual, EU plug, UK plug, CHN plug, US/Japan plug.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty