Would you like to log in again?
COMBI HEAD ACCESSORY DW 96 SAMPLES
Supplier: LIFE TECHNOLOGIES CORP
Combi head^ KingFisher* Apex Combi head, Volume (Metric) Processing: KF 96 plate 15-200 ul, 96 DW plate 15-1000 ul, 96 storage tube 30-200 ul, Plate Type: 96 KF, 96 DW, 96 storage tube, Particle Size (Metric): ca. > 1 um, 96 Samples
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COMBI HEAD ACCESSORY DW 96 SAMPLES 76417-648 24079920 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty