Would you like to log in again?
Polypropylene Vials, Starplex®
Supplier: Starplex Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V112-YNL V8084-S V103-NL V102-NL V103-NL V8093 V102 V8095 V102-NL V8093-S V8094 V8084 V102-POUCH
76446-930CS 175.42 USD InStock
CA73240-127 76446-930 CA73240-129 14216-268 CA73240-133 CA10861-530 14216-270 CA11006-064 76417-694 CA73270-066 CA66410-024 76417-696 11007-280
Polypropylene Vials, Starplex®
Vials
These vials are made of crack-resistant medical grade polypropylene.
  • With or without screw caps
  • Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty