Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GS™ COVID-19 Real-Time PCR Kit, Genosensor

Supplier: Genosensor
2702-94 2702-22
76417-830EA 991.81 USD InStock
76417-830 76417-832
GS™ COVID-19 Real-Time PCR Kit, Genosensor
Microbiology Test Systems PCR Tests
Quick and reliable solutions for 2019 coronavirus disease(COVID-19) and SARS-CoV-2.

  • Accuracy: Assay 3 target genes unique to SARS-CoV-2 having higher specificity and exhibiting lower risk for mutation
  • Specificity: Targeted specificity to 100% of currently available complete genomes for SARS-CoV-2.
  • Simplicity: Ready to use all-in-one master mix reagents and controls minimize variability
  • Throughput: Two kit package options of viral detection for up to 94 specimens
  • High quality, high performance and reliability

The GS™ COVID-19 RT-PCR KIT provides accurate, reproducible, high-quality results for clinical decision-making for patients with suspected COVID-19 (coronavirus) infection. The GS™ COVID-19 RT-PCR KIT performs a qualitative molecular assay that allows the detection of SARS-COV-2 nucleic acids in samples from patients who meet COVID-19 clinical and/or epidemiological criteria.

The GS™ COVID-19 RT-PCR KIT is a real-time reverse transcription polymerase chain reaction (RT-PCR) test which detects three genes from the SARS-CoV-2 virus from clinical nasopharyngeal swab specimens. In addition, the GS™ COVID-19 RT-PCR KIT also includes an internal positive control gene (GUSB) that serves as an extraction, reverse transcription, and PCR amplification control for each sample to minimize false negative results.

The GS™ COVID-19 RT-PCR KIT runs on the Applied Biosystems™ 7500 Fast Dx Real-Time PCR Instrument. Kits are available for the 96 well plate (22 clinical samples) and 384 well plate (94 clinical samples) and provides test results in less than 90 minutes.

Certifications: For more up-to-date information on the current coronavirus outbreak, visit the WHO or CDC websites. This test has not been FDA cleared or approved; This test has been authorized by FDA under an EUA for use by authorized laboratories; This test has been authorized only for the detection of nucleic acid from SARS-CoV-2, not for any other viruses or pathogens; and, This test is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Act, 21 U.S.C. § 360bbb-3(b)(1), unless the authorization is terminated or revoked sooner.

Caution: For use under the Emergency Use Authorization (EUA) only. For in vitro diagnostic use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty