Would you like to log in again?
Human Recombinant BMPR-1A (from CHO cells)
Supplier: Enzo Life Sciences
PRT260-0020
76418-038EA 620.7 USD InStock
76418-038
Human Recombinant BMPR-1A (from CHO cells)
Proteins and Peptides
Produced in genetically modified CHO cells. Contains a C-terminus linker (IEGRMD) and an Fc tag (Human IgG1).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty