Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rimadyl® MD Tablets for Rodents

Supplier: Bio-Serv
Rimadyl®
MD150-2-C MD150-2-C SMD150-2 MD150-2 MD150-2-25 MD150-2 SMD150-2 MD150-2-25
89067-456PK 104.68 USD InStock
89067-456 CA76628-362 CA89067-456 CA77506-112 77506-112 76419-510 CA76419-510 76628-362
Rimadyl® MD Tablets for Rodents
Animal Supplies Animal Feed
Rimadyl (carprofen) is an effective anti-inflammatory drug used in a variety of species including rodents. Ideal pain-free alternative to stressful and laborious post-op injections.

  • Highly palatable, nutritionally complete, grain-based, bacon-flavored tablets are easy to administer
  • Simply place tablets on cage floor for easy access. More convenient and less risk of dehydration compared to using medicated water
  • Cold tableting process eliminates drug degradation
  • MD's can be customized to meet specific requirements
  • Each 5 g tablet is 1" diameter ×0.25" high
  • Shelf life: 6 months, cool dry storage

Ingredients: Ground corn, banana flakes, dehulled soybean meal, corn gluten meal, ground wheat, corn gluten feed, fish meal, dehydrated alfalfa meal, casein, dried whey, sucrose, fructose, dextrose, dried beet pulp, soybean oil, porcine animal fat (preserved with BHA), dried brewers yeast, mineral mix, vitamin mix, magnesium stearate, calcium silicate, bacon flavor, DL-methionine, choline chloride, sodium propionate, ascorbic acid, kaolin.

Mouse average dose: One 5 g tablet/20 g of body weight. Rat average dose: One 5 g tablet/100 g of body weight.

Caloric value: 3.43 kcal/g, 17.2 kcal/tablet
Nutritional ash content: 6.7%
Nutritional carbohydrate content: 55.0%
Nutritional fat content: 4.0%
Nutritional fibre content: 4.0%
Nutritional moisture content: 5.0%
Nutritional protein content: 21.1%

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty