Would you like to log in again?
Primate MD's™ Anti-Inflammatory
Supplier: Bio-Serv
MD's™
MD650-90 MD650-90 F07096 F07096
76419-512PK 78.28 USD InStock
CA76285-256 CA76419-512 76419-512 76285-256
Primate MD's™ Anti-Inflammatory
Animal Supplies Animal Feed
Chewable, flavored, medicated tablet replaces more stressful dosing methods.
  • Tablet consistency ensures accurate dosing
  • Scored 5 gm tablet for flexible dose administration
  • Cold tableting process eliminates drug degradation
  • No heat or water used in processing
  • Highly palatable sugar-based formula with excellent drug masking capabilities
  • Reduces time, labor and stress compared to traditional dosing methods
  • MD's can be customized to meet specific requirements
  • Shelf life: 6 to 24 months as dictated by the drug expiration date
Base Meal: Sucrose, dextrose, mineral oil, artificial and natural flavors, calcium silicate, magnesium stearate.
.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty