Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Veggie-Bites™ Guinea Pig Enrichment Treats, Certified

Supplier: Bio-Serv
Veggie-Bites™
F5158-1 S5158-1 F5158-1 F5158 F5158
76419-518BG 23.19 USD InStock
76419-518 CA89067-444 CA76419-518 76406-494 89067-444
Veggie-Bites™ Guinea Pig Enrichment Treats, Certified
Animal Supplies Animal Feed
An all-natural, crunchy vegetable treat that is a medley of dehydrated sliced carrots, puffed peas and whole kernel corn.

  • Less labor intensive and more sanitary than perishable vegetables
  • Feed as a training reward or foraging treat
  • Nutritionally assayed
  • Shelf life: 12 months, cool dry storage

Ingredients: Dehydrated carrots, puffed peas, dried whole kernel corn.

Caloric value: 3.44 kcal/gm
Nutritional ash content: 2.0%
Nutritional carbohydrate content: 67.0%
Nutritional fat content: 4.0%
Nutritional fibre content: 5.0%
Nutritional moisture content: <12%
Nutritional protein content: 10.0%

Certifications: Certified (Contaminant screened).

Ordering information: Product is no longer gamma irradiated. It is now Certified. If gamma irradiation is required, please contact customer service for pricing and lead time.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty