Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Feeding Needles for PO Gavage Dosing in Small Animals, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
FN15-100M FN17-52M FN15-78M FN20-38 FN17-85 FN18-50M FN18-38 FN20-38M FN17-85M FN18-38M FN17-52 FN15-78 FN20-30
76419-526PK 429.3 USD InStock
76419-526 76419-536 76419-528 76419-538 76419-522 76419-544 76419-532 76419-546 76419-524 76419-534 76419-540 76419-542 76419-530
Feeding Needles for PO Gavage Dosing in Small Animals, SAI Infusion Technologies
Animal Supplies Animal Feeding Needles
Flexible PTFE feeding needles are designed to reduce the risk of trauma and bite-through, providing a safe, low-cost, disposable, solution for oral gavage. The affordable feeding tubes boast distinct features, including a soft silicone tip, a smooth plastic PTFE shaft and an optional, bite-resistant, stainless steel bite-guard for maximum durability when oral dosing.
  • Supplied sterile
  • Affordable and durable
  • Bulb-shaped silicone tip minimizes trauma, prevents tracheal dosing, and eliminates dosing-related perforations of the esophagus
  • Plastic construction is compatible with most vehicles and compounds
  • Disposable needles eliminate the risk of carry-over
Flexible feeding needles (gavage needles) have countless laboratory applications. Not only for use when oral dosing small animals, these needles may be used for a variety of surgical and delivery procedures. These needles provide a smooth, burr-free silicone tip which prevents inadvertent damage to the esophagus caused by inferior needles. Add the optional stainless steel bite-guard to combine the flexibility of a plastic needle with the durability of stainless steel.
Optional 1 cm stainless steel bite-guard near the hub is designed to prevent damage from the animal's incisors during oral dosing.
Packaging: Needles are packaged 100/box (20 packs of 5).
Caution: Designed for animal research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty