Would you like to log in again?
VWR BAG AUTOCLAVE BIO-HAZARD PP ST CS50
Supplier: VWR International
VWR* Bag^ VWR* Pre Sterilized Autoclavable Biohazard Bag, Material: polypropylene, Color: red, and printed with universal Biohazard symbol, and conforms to all Federal and State regulations worldwide, Includes a temperature indicator, 3 mil, Dimension: 37x48in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BAG AUTOCLAVE BIO-HAZARD PP ST CS50 76419-582 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty