Would you like to log in again?
Free Standing Biohazard Shield, TrippNT
Supplier: TrippNT
53408
76419-596EA 343.94 USD InStock
76419-596
Free Standing Biohazard Shield, TrippNT
Face Shields
TrippNT's free standing biohazard shield is made from high-quality acrylic and HDPE to help prevent cross-contamination at work stations.
  • Ships flat with easy snap-in assembly
  • Works as biohazard shield or bench divider
  • Clear acrylic construction
  • Can be cleaned easily with wipes or mild detergents
The Biohazard shield can be easily disinfected with isopropyl alcohol or mild detergents, preventing cross-contamination between work stations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty