Would you like to log in again?
SP Bel-Art ProCulture Petri Dish Racks, 100 mm, Bel-Art Products, a Part of SP
Supplier: Bel-Art Products, a part of SP
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18982-0009 F18982-0014 F18982-0409 F18982-0109 F18982-0209 F18982-0314 F18982-0309
76419-628EA 245.25 USD InStock
76419-628 76419-636 76419-638 76419-626 76419-632 76419-634 76419-630
SP Bel-Art ProCulture Petri Dish Racks, 100 mm, Bel-Art Products, a Part of SP
Racks Petri Dish Racks
Increase workspace and efficiently transport large quantities of Petri dishes.
  • Silicone holding strips grip each Petri dish; easily insert and remove groups of dishes without disturbing upper or lower dishes
  • Side labels provide space for critical sample identification
  • Lightweight, chemical resistant polycarbonate
  • Autoclavable at 121 °C (250 °F)
Available in four colors and two sizes; the 24.1 cm (9½") high size holds forty-eight 100 mm Petri dishes, and the 36.8 cm (14½") high size holds eighty-four 100 mm Petri dishes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty