Would you like to log in again?
Vivaspin® Turbo 15 RC, Sartorius
Supplier: Sartorius
Vivaspin®
VS15TR11 VS15TR12 VS15TR31 VS15TR01 VS15TR01 VS15TR21 VS15TR21 VS15TR22 VS15TR32 VS15TR02 VS15TR12 VS15TR02 VS15TR41 VS15TR31 VS15TR22 VS15TR11 VS15TR42 VS15TR32 VS15TR42
76420-030PK 185.4 USD InStock
CA76416-004 CA76416-002 CA76416-006 76420-030 CA76415-996 76420-040 CA76415-998 76420-034 CA76415-992 76420-044 76420-032 CA76415-994 76420-042 76420-038 76420-026 76420-036 CA76416-000 CA76416-010 76420-028
Vivaspin® Turbo 15 RC, Sartorius
Filters Centrifugal Filters
Vivaspin® Turbo 15 Regenerated Cellulose membrane, ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4 to 15 ml sample volumes.
  • Unique optimal membrane choice (PES and RC)
  • Complete recoveries
  • Fastest concentrations
  • Highest chemical resistance
  • Easy retentate collection
  • Graduated and markable
Vivaspin® Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs.
The Vivaspin® Turbo 15 RC concentrators are the regenerated cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 µl.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty