Would you like to log in again?
CRYO RACK 1272XX F/48 CRYO TUBES CS15
Supplier: Greiner Bio-One
Cryo rack^ Cryo rack, 1272XX, pre barcoded, black, high profile, code 128 with check digit, s1 centered, human readable number include check digit, data matrix code on rack bottom, for 48 cryo tubes
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYO RACK 1272XX F/48 CRYO TUBES CS15 76420-050 803270-2DG Case of 15 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty