Would you like to log in again?
Fluorescent Viewing Goggles, Noir medicals
Supplier: NOIR MEDICAL
U60
76420-054EA 39.86 USD InStock
76420-054
Fluorescent Viewing Goggles, Noir medicals
Educational Materials Forensics Educational Materials Forensics Supplies
Necessary protection when operating forensic light source.
Completely shield your eyes from UV A and UV B rays with these flexible orange viewing goggles. Clearly see latent prints and other evidence fluorescing with our wrap around goggles that meet ANSI standards and comfortably fit over prescription glasses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty