Would you like to log in again?
Axygen® Automation Tips for Agilent®/Velocity11 VPrep® and Bravo Workstations, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
VT-255-L-R-S VT-250-R-LAB-S VT-384-5-L-R VT-384-70-L-R-S VT-384-10UL-R VT-250-R-LAB VTF-384-50UL-R-S VT-255-L-R VT-384-10UL-R-S VTF-205-L-R-S VT-384-31UL-R VT-384-31-L-R VT-384M2-70UL-R VT-384-31-L-R-S VTF-384-15-L-R-S VTF-384-50-L-R-S VT-384-70-L-R VT-384-10-L-R VTF-205-R-S VT-384-70UL-R VT-384-5-L-R-S VT-250-R-S VT-50-R VT-255-R-S VT-30UL-R VTF-165-R-S VT-384-31UL-R-S VT-250-L-R VT-384-70UL-R-S VT-255-R VTF-20-R-S VT-20-R VT-250-R VTF-384-15UL-R-S
89096-108CS 4396.6 USD InStock
89096-108 89425-680 89025-856 89096-100 89425-682 89425-686 89425-688 76420-062 89096-082 89425-678 89096-084 76420-060 89096-062 76420-088 89096-064 76420-064 89413-110 10799-771 CA10790-704 89096-080 89125-538 89124-992 89124-994 89096-078 89425-670 89425-672 89096-114 89425-674 89096-116 89425-676 76420-056 89096-076 89096-110 76420-058
Axygen® Automation Tips for Agilent®/Velocity11 VPrep® and Bravo Workstations, Corning
Pipette Tips
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Maxymum Recovery® version available in most tip styles
  • Filtered version available in most tip styles to prevent aerosol and liquid contamination
Axygen® offers a comprehensive line of tips for Agilent®/Velocity11 automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.Available in nonsterile, sterile, non-filtered, filtered, and wide-bore formats. Also available in Maxymum Recovery® to further improve accuracy and precision while reducing sample loss. Clear polypropylene tips are available in 96-well and 384-well format designed for Agilent®/Velocity VPrep® and Bravo 96LT, 8LT, 384ST, 96ST and 16ST heads.
Certifications: Tips are certified RNase-/DNase-free, and nonpyrogenic.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty