Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Entris® II Advanced Precision Laboratory Balances, Sartorius
Supplier: Sartorius
Entris® II
BCA10201I-1S BCA2201-1S BCA2201I-1S BCA2202I-1S BCA12201I-1S BCA423-1S BCA822I-1S BCA1202I-1S BCA822-1S BCA5201-1S BCA623-1S BCA2202-1S BCA6202-1S BCA323I-1S BCA8201I-1S BCA4202-1S BCA5201I-1S BCA323-1S BCA2201I-1S BCA1202I-1S BCA8201-1S BCA10201-1S BCA623-1S BCA1203I-1S BCA10201I-1S BCA223I-1S BCA6202I-1S BCA2202I-1S BCA6202I-1S BCA623I-1S BCA423-1S BCA5201-1S BCA822I-1S BCA12201-1S BCA3202I-1S BCA1203I-1S BCA12201-1S BCA223-1S BCA223-1S BCA223I-1S BCA423I-1S BCA10201-1S BCA323I-1S BCA3202-1S BCA8201I-1S BCA2202-1S BCA12201I-1S BCA1202-1S BCA3202I-1S BCA1202-1S BCA8201-1S BCA623I-1S BCA5201I-1S BCA822-1S BCA3202-1S BCA4202I-1S BCA2201-1S BCA4202I-1S BCA4202-1S BCA323-1S BCA423I-1S BCA6202-1S
76420-162EA 4212.51 USD InStock
76420-162 76420-160 76420-122 76420-166 CA76420-130 CA76420-174 76420-164 CA76420-172 76420-126 CA76420-178 76420-124 CA76420-176 CA76420-132 CA76420-138 76420-128 76420-172 CA76420-142 76420-132 76420-176 CA76420-140 76420-174 76420-130 CA76420-144 76420-178 CA76420-106 CA76420-148 CA76420-104 76420-138 CA76420-108 76420-140 76420-144 CA76420-152 76420-142 CA76420-150 76420-104 76420-148 CA76420-156 CA76420-112 CA76420-110 CA76420-154 76420-108 CA76420-116 76420-106 CA76420-158 CA76420-114 CA76420-160 76420-150 CA76420-164 76420-110 76420-154 CA76420-162 76420-152 CA76420-124 76420-114 76420-158 CA76420-166 CA76420-122 76420-156 76420-112 CA76420-128 CA76420-126 76420-116
Entris® II Advanced Precision Laboratory Balances, Sartorius
Balances and Scales
Entris® II Basic Advanced precision balances feature isoCAL, intuitive graphic touch display and 13 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard-wearing PBT, stainless steel and glass. Integrated protection systems increase reliability of weighing results: 3 configurable levels determine valid weighing data and ensure only valid data is transferred to external devices.
  • Fast stabilisation time thanks to state-of-the-art weighing sensors
  • Best repeatability with rectangular weighing pan
  • Overload protection, rugged design, weighs up to a pre-set amount
  • Guaranteed reliability with the self-test ‘@start’
  • Choice of models with internal or external calibration
  • CalAudit Trail provides gapless documentation with automatic documentation of calibration processes, calibration trigger and date and time stamp
  • Hybrid screen gives excellent readability and use. Combines intuitive, wear resistant LED and touch technology provides an easy and clear structured user interface
  • Password protection ensures secure operation, avoids unintentional changes. User management controls access to balance settings
  • Built-in real time level support simplifies balance levelling
The models with 0.1 and 1 mg readability have a draft shield, this minimises weighing errors caused by electrostatically charged samples using specially coated glass parts. In models up to 620 g the weighing chamber gives the highest usable weighing chamber height easily accommodating even large containers, ensuring convenient access to the weighing pan. The sliding and top doors can be can easily be removed for cleaning. The models with 10 or 100 mg readability do not have draft shields.
Choice of models with isoCAL (internal calibration and adjustment) for accurate weighing results. Optimal accuracy and operating convenience using fully automated internal temperature and time-controlled calibration and adjustment feature. Assures (SOP) compliant operation with self-notification if calibration is outside the normal range. The balance easily adapts to the ambient conditions with just one click on the home screen. An internal electronic level sensor continuously monitors for proper levelling, alarm messaging when it is not levelled and interactive user guidance.
Plug-and-play convenience - automatically detects Sartorius accessories (e.g. printer, second display). Real "PC-direct feature" for easy connection to a PC to transfer weighing data directly into spreadsheets or documents such as Microsoft® Excel or Word.
The models have built-in applications with GLP/GMP compliant printout/data printout. Functions include: weighing/dosing, counting, percentage weighing, mixing/net total, components/totalizing, dynamic weighing, calculation/fee factor, density determination, statistics, peak hold, check weighing, mass unit conversion, underfloor weighing for bigger samples and pipette smart test.
Entris® II Basic Advanced balances have 13 built-in applications with GLP / GMP compliant printout / data output. All models have 13 built-in languages for international operation: English, German, French, Italian, Spanish, Portuguese, Russian, Chinese, Japanese, Korean, Turkish, Hungarian and Polish. The series has data output for dynamic weighing applications and configurable time interval for data output and an RS232 9-pin interface and a RS232 port. Two USB Type C interfaces enable multiple connections to external devices, data can also be transferred to a USB stick. Entris II allows individualized printouts for up to 6 unique use profiles. The models have an anti-theft lock.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty