Would you like to log in again?
SLEEVE CLASS 5-8 STERILE 18IN CS 50 PAIR
Supplier: CHEMLAND-LAKELAND INDUS
Sleeve^ Sleeve, Clean Sterile, has bound seams to prevent break through and strike through, Thumb loops on elastic wrist keep the sleeve anchored in place to prevent accidental exposure, has a bacterial and particle filtration efficiency of 99.999%, Fluid Repellant, ISO Class 5 - 8, Length: 18in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SLEEVE CLASS 5-8 STERILE 18IN CS 50 PAIR 76420-182 CTL850CSP-18 Case of 50 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty