Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Bottle Top Dispensers, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Corning®
6843 6844 6840 6841 6845 6842
76420-194CS 393.94 USD InStock
76420-194 76420-192 76420-198 76420-196 76420-200 76420-190
Corning® Bottle Top Dispensers, Corning
Bottle Top Dispensers
Corning® bottle top dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent. Depending on the model, they can accommodate volumes from 0.25 to 100 ml.

  • Accurate and reproducible volume setting
  • Fully autoclavable for protection against contamination
  • Rotating valve block for convenient dispensing
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system for user safety at work
  • Easy in-lab calibration with built-in recalibration mechanism

In-lab calibration tool and set of four thread adapters included with every product.

Certifications: Meets ISO 8655 requirements.

Delivery information: Dispensers up to 10 ml are delivered with adapter sizes: A25, A28, A40, A45. Dispensers up to 25, 50, and 100 ml are delivered with adapter sizes: A25, A28, A38, A45/32.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty